Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000430968.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000430968.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000430968.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000430968.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000430968.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000430968.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000449869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000449869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000449869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000458007.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000458007.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000458007.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000458007.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000467775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000467775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000467775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
ENST00000467775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013243.hg.1 |
TC02000894.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013243.hg.1 |
ENST00000280098.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013243.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200166387.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166387.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166387.hg.1 |
ENST00000430968.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166387.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200166387.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166387.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166387.hg.1 |
ENST00000430968.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166387.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200166387.hg.1 |
TC0200009487.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000280098.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000420679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000430968.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000430968.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000430968.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000430968.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000430968.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000430968.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000449869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000449869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000449869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000458007.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000458007.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000458007.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000458007.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000467775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000467775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000467775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
ENST00000467775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009487.hg.1 |
NM_001001664.2 |
Refseq |