Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000443151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000443151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000443151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000443151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000443151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000443151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000443151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000469980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000469980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000469980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000469980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000472667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000472667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000472667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000472667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000472667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000472667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000475571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000475571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000475571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000475571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000475571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
ENST00000475571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02014661.hg.1 |
TC02005001.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02014661.hg.1 |
ENST00000295240.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02014661.hg.1 |
ENST00000392663.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02014661.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200223982.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200223982.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200223982.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200223982.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200223982.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200223982.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200223982.hg.1 |
TC0200016559.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000295240.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000392663.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000443151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000443151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000443151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000443151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000443151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000443151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000443151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000469980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000469980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000469980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000469980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000472667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000472667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000472667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000472667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000472667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000472667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000475571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000475571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000475571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000475571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000475571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
ENST00000475571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016559.hg.1 |
NM_152384.2 |
Refseq |