Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044694.hg.1 |
TC01002866.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044694.hg.1 |
ENST00000370360.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044694.hg.1 |
ENST00000534881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044694.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000471465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000471465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000471465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000471465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000471465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000487911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000487911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000487911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000487911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000471465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000471465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000471465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000471465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000471465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000487911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000487911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000487911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000487911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
ENST00000495852.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014921.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211152.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211152.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211152.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211152.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211152.hg.1 |
ENST00000370360.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211152.hg.1 |
ENST00000463560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211152.hg.1 |
ENST00000495106.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211152.hg.1 |
NM_053274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211152.hg.1 |
TC0100014921.hg.1 |
Clariom D TC |