Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008481.hg.1 |
ENST00000217195.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008481.hg.1 |
ENST00000379765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008481.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008481.hg.1 |
ENST00000399672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008481.hg.1 |
ENST00000487464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008481.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008481.hg.1 |
TC20000576.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008486.hg.1 |
ENST00000217195.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008486.hg.1 |
ENST00000379765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008486.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008486.hg.1 |
ENST00000399672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008486.hg.1 |
ENST00000487464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008486.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008486.hg.1 |
TC20000576.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000399683.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000399683.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000399683.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
ENST00000399683.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008230.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008484.hg.1 |
ENST00000217195.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008484.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008484.hg.1 |
ENST00000399672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008484.hg.1 |
ENST00000487464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008484.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008484.hg.1 |
TC20000576.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000158103.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158103.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158103.hg.1 |
TC2000008230.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000158103.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158103.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158089.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158089.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158089.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158089.hg.1 |
ENST00000399683.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158089.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158089.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158089.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158089.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158089.hg.1 |
TC2000008230.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000158089.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158089.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158089.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158089.hg.1 |
ENST00000399683.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158089.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158089.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158089.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158089.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000399683.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000399683.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000399683.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
ENST00000399683.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008230.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158096.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158096.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158096.hg.1 |
TC2000008230.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000158096.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158096.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158090.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158090.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158090.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158090.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158090.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158090.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158090.hg.1 |
NR_047675.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158090.hg.1 |
TC2000008230.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000158090.hg.1 |
ENST00000217195.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158090.hg.1 |
ENST00000379772.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158090.hg.1 |
ENST00000399672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158090.hg.1 |
NM_001039140.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158090.hg.1 |
NM_001258429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158090.hg.1 |
NM_001258430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158090.hg.1 |
NR_047675.1 |
Refseq |