Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007659.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007659.hg.1 |
NM_001008778.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007659.hg.1 |
TC11000619.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008004.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008004.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008004.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008004.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008004.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008004.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008004.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008004.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008004.hg.1 |
NM_001008778.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008004.hg.1 |
NM_001008778.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008004.hg.1 |
NM_001008778.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008004.hg.1 |
NM_001008778.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008004.hg.1 |
NM_001008778.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008004.hg.1 |
NM_001008778.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008004.hg.1 |
NM_001008778.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157614.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157614.hg.1 |
NM_001008778.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157614.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157614.hg.1 |
NM_001008778.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157614.hg.1 |
TC1100008004.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100008004.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008004.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008004.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008004.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008004.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008004.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008004.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008004.hg.1 |
ENST00000377185.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008004.hg.1 |
NM_001008778.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008004.hg.1 |
NM_001008778.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008004.hg.1 |
NM_001008778.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008004.hg.1 |
NM_001008778.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008004.hg.1 |
NM_001008778.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008004.hg.1 |
NM_001008778.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008004.hg.1 |
NM_001008778.2 |
Refseq |