Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000427750.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000427750.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000427750.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000427750.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000427750.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000427750.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000427750.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000427750.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000527207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000527207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000527207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000527207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000530920.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000530920.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020733.hg.1 |
ENST00000358252.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020733.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020733.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020733.hg.1 |
TC11001787.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100182211.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182211.hg.1 |
ENST00000527207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182211.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182211.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182211.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182211.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182211.hg.1 |
ENST00000527207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182211.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182211.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182211.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182211.hg.1 |
TC1100010885.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000358252.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000427750.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000427750.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000427750.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000427750.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000427750.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000427750.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000427750.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000427750.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000527207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000527207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000527207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000527207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000528882.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000530920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000530920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
ENST00000532437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010885.hg.1 |
NM_033396.2 |
Refseq |