Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs01125663_m1
See other RBFOX1 GE Assays ›
Gene Symbol:
RBFOX1
Gene Name:
RNA binding protein, fox-1 homolog 1
Gene Aliases:
2BP1, A2BP1, FOX-1, FOX1, HRNBP1
Chromosome Location:
Chr.16: 5239749 - 7713343 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs01125663_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: RBFOX1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001142333.1
NP_001135805.1
12 - 13
1843
73
NM_001142334.1
NP_001135806.1
11 - 12
1156
73
NM_001308117.1
NP_001295046.1
13 - 14
1261
73
NM_018723.3
NP_061193.2
13 - 14
1924
73
XM_011522546.2
XP_011520848.1
10 - 11
1153
73
XM_011522547.2
XP_011520849.1
10 - 11
1153
73
XM_011522548.2
XP_011520850.1
9 - 10
1072
73
XM_017023319.1
XP_016878808.1
15 - 16
1466
73
XM_017023322.1
XP_016878811.1
12 - 13
3017
73
XM_017023323.1
XP_016878812.1
10 - 11
1174
73
XM_017023324.1
XP_016878813.1
13 - 14
1955
73
XM_017023326.1
XP_016878815.1
13 - 14
2046
73
XM_017023327.1
XP_016878816.1
10 - 11
1174
73
XM_017023328.1
XP_016878817.1
13 - 14
1955
73
XM_017023329.1
XP_016878818.1
12 - 13
1874
73
XM_017023330.1
XP_016878819.1
12 - 13
2494
73
XM_017023332.1
XP_016878821.1
10 - 11
1174
73
XM_017023333.1
XP_016878822.1
12 - 13
1056
73
XM_017023334.1
XP_016878823.1
9 - 10
1093
73
XM_017023335.1
XP_016878824.1
13 - 14
2046
73
XM_017023336.1
XP_016878825.1
12 - 13
1056
73
XM_017023337.1
XP_016878826.1
9 - 10
1093
73
XM_017023338.1
XP_016878827.1
11 - 12
975
73
XM_017023339.1
XP_016878828.1
10 - 11
1200
73
XM_017023340.1
XP_016878829.1
11 - 12
963
73
XM_017023341.1
XP_016878830.1
12 - 13
1965
73
XM_017023342.1
XP_016878831.1
11 - 12
975
73
XM_017023343.1
XP_016878832.1
9 - 10
1122
73
GenBank mRNA
AF107203.1
-
13 - 14
1922
73
AF448859.1
-
12 - 13
1841
73
AK001027.1
-
11 - 12
1156
73
AK293951.1
-
13 - 14
1261
73
AK299664.1
-
10 - 11
834
73
KJ534926.1
-
10 - 11
996
73
KJ535000.1
-
10 - 11
996
73
KJ535001.1
-
9 - 10
915
73
Back To Top
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact
Tech Support
if you need more information.
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene RBFOX1
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of alternative mRNA splicing, via spliceosome
mRNA processing
nervous system development
RNA splicing
RNA transport
nucleotide binding
RNA binding
mRNA binding
protein binding
protein C-terminus binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-bn9m5:80/10.239.167.51:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0