Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000471984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000471984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000471984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000471984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000490058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000490058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
ENST00000490058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007620.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007620.hg.1 |
ENST00000378302.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007620.hg.1 |
ENST00000546015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007620.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007620.hg.1 |
TC13000661.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000471984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000471984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000471984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000471984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000490058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000490058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
ENST00000490058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008918.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300160213.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300160213.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300160213.hg.1 |
ENST00000490058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300160213.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300160213.hg.1 |
ENST00000258646.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300160213.hg.1 |
ENST00000378302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300160213.hg.1 |
ENST00000490058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300160213.hg.1 |
NM_018191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300160213.hg.1 |
TC1300008918.hg.1 |
Clariom D TC |