Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020061.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020061.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020061.hg.1 |
TC12001527.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020062.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020062.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020062.hg.1 |
TC12001527.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200182052.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182052.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182052.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182052.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182052.hg.1 |
TC1200010761.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200182053.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182053.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182053.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182053.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182053.hg.1 |
TC1200010761.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
ENST00000267119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010761.hg.1 |
NM_033448.2 |
Refseq |