Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006900.hg.1 |
ENST00000376221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006900.hg.1 |
ENST00000376221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006900.hg.1 |
ENST00000376221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006900.hg.1 |
ENST00000376221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006900.hg.1 |
ENST00000376221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006900.hg.1 |
NM_001013630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006900.hg.1 |
NM_001013630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006900.hg.1 |
NM_001013630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006900.hg.1 |
NM_001013630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006900.hg.1 |
NM_001013630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002714.hg.1 |
TC01000159.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002714.hg.1 |
ENST00000376221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002714.hg.1 |
NM_001013630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002715.hg.1 |
TC01000159.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002715.hg.1 |
ENST00000376221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002715.hg.1 |
NM_001013630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148109.hg.1 |
ENST00000376221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148109.hg.1 |
NM_001013630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148109.hg.1 |
ENST00000376221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148109.hg.1 |
NM_001013630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148109.hg.1 |
TC0100006900.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100006900.hg.1 |
ENST00000376221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006900.hg.1 |
ENST00000376221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006900.hg.1 |
ENST00000376221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006900.hg.1 |
ENST00000376221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006900.hg.1 |
ENST00000376221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006900.hg.1 |
NM_001013630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006900.hg.1 |
NM_001013630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006900.hg.1 |
NM_001013630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006900.hg.1 |
NM_001013630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006900.hg.1 |
NM_001013630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148110.hg.1 |
ENST00000376221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148110.hg.1 |
NM_001013630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148110.hg.1 |
ENST00000376221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148110.hg.1 |
NM_001013630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148110.hg.1 |
TC0100006900.hg.1 |
Clariom D TC |