Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
8119388 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8119389 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8119386 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
230236_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11744459_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11744459_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006251.hg.1 |
TC06000559.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006251.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006251.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006251.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006252.hg.1 |
TC06000559.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006252.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006252.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006253.hg.1 |
TC06000559.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006253.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006253.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006253.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006254.hg.1 |
TC06000559.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006254.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006254.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006254.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006254.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006254.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155209.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155209.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155209.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155209.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155209.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155209.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155209.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155209.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155209.hg.1 |
TC0600007969.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600155214.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155214.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155214.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155214.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155214.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155214.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155214.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155214.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155214.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155214.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155214.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155214.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155214.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155214.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155214.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155214.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155214.hg.1 |
TC0600007969.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600155213.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155213.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155213.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155213.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155213.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155213.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155213.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155213.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155213.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155213.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155213.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155213.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155213.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155213.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155213.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155213.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155213.hg.1 |
TC0600007969.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155210.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155210.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155210.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155210.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155210.hg.1 |
TC0600007969.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600155211.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155211.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155211.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155211.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155211.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155211.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155211.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155211.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155211.hg.1 |
TC0600007969.hg.1 |
Clariom D TC |
Human Genome U133 Plus 2.0 Array |
230236_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
230236_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230236_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230236_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230236_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230236_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230236_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230236_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230236_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230236_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230236_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230236_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230236_at9 |
- |
- |
Human Genome U95 Set |
58971_at |
- |
- |
Human Genome U95 Set |
58971_PM_at |
- |
- |
Human Genome U133 Set |
230236_PM_at |
- |
- |
Human Genome U133 Set |
230236_at |
- |
- |