Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007888.hg.1 |
ENST00000217260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007888.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007888.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007888.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007888.hg.1 |
TC20000547.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007886.hg.1 |
ENST00000217260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007886.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007886.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007886.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007886.hg.1 |
TC20000547.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008146.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007887.hg.1 |
ENST00000217260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007887.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007887.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007887.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007887.hg.1 |
TC20000547.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008146.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157213.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157213.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157213.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157213.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157213.hg.1 |
TC2000008146.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000157213.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157213.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157213.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157213.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157215.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157215.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157215.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157215.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157215.hg.1 |
TC2000008146.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000157215.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157215.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157215.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157215.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157214.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157214.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157214.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157214.hg.1 |
NM_001040007.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157214.hg.1 |
TC2000008146.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000157214.hg.1 |
ENST00000217260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157214.hg.1 |
ENST00000400634.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157214.hg.1 |
NM_001029871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157214.hg.1 |
NM_001040007.2 |
Refseq |