Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009507.hg.1 |
ENST00000595655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009507.hg.1 |
ENST00000595655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397981.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397981.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397981.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397981.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397981.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397981.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397981.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397981.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397981.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397981.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397981.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397981.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397981.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397981.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397981.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397981.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397981.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000397983.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000465324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000465324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000465324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000465324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000465324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000468058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000468058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000468058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000468058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000468058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000468058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000468058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000468058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000468058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000468058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000468058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000468058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000468058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000468058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000468058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000468058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000468058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000475022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000475022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000475022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000475022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000475022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000494348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000494348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000494348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000494348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000494348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000494348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000494348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000494348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000498118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000498118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
ENST00000498118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297555.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_001297556.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_145185.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_145185.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_145185.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_145185.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_145185.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_145185.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_145185.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_145185.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_145185.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_145185.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_145185.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_145185.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_145185.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_145185.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_145185.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_145185.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_145185.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006864.hg.1 |
NM_145185.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009507.hg.1 |
ENST00000595655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009507.hg.1 |
ENST00000595655.1 |
Ensembl |