Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018334.hg.1 |
ENST00000262809.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018334.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018334.hg.1 |
TC19001290.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018333.hg.1 |
ENST00000262809.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018333.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018333.hg.1 |
TC19001290.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000596915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000596915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000596915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000608165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000608165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000608165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000608165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000610152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000610152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000610152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000610152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
ENST00000610152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000596915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000596915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000596915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000608165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000608165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000608165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000608165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000610152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000610152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000610152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000610152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
ENST00000610152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010017.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900182303.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182303.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182303.hg.1 |
ENST00000610152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182303.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900182303.hg.1 |
TC1900010017.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900182303.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182303.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182303.hg.1 |
ENST00000610152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182303.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900182305.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182305.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182305.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182305.hg.1 |
ENST00000610152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182305.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900182305.hg.1 |
TC1900010017.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900182305.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182305.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182305.hg.1 |
ENST00000596124.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182305.hg.1 |
ENST00000610152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182305.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900182301.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182301.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182301.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900182301.hg.1 |
TC1900010017.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900182301.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182301.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182301.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900182302.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182302.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182302.hg.1 |
ENST00000610152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182302.hg.1 |
NM_006532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900182302.hg.1 |
TC1900010017.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900182302.hg.1 |
ENST00000262809.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182302.hg.1 |
ENST00000594635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182302.hg.1 |
ENST00000610152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182302.hg.1 |
NM_006532.3 |
Refseq |