Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003986.hg.1 |
ENST00000262865.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003986.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003986.hg.1 |
TC20000281.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003987.hg.1 |
ENST00000262865.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003987.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003987.hg.1 |
TC20000281.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000489039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000489039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000489039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000489039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000490381.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000490381.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000490381.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000490381.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
ENST00000490381.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169766.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169766.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169766.hg.1 |
TC2000009918.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000169766.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169766.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169765.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169765.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169765.hg.1 |
TC2000009918.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000169765.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169765.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000262865.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000417318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000489039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000489039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000489039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000489039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000489102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000490381.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000490381.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000490381.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000490381.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
ENST00000490381.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009918.hg.1 |
NM_001725.2 |
Refseq |