Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
ENST00000269844.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
ENST00000398548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
ENST00000422911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
ENST00000433067.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
ENST00000441787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
ENST00000447016.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
ENST00000447207.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
ENST00000449395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
NM_001040424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
NM_001282934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
NM_022115.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
NR_104257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
NR_104258.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
NR_104260.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
TC2100008227.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
ENST00000269844.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
ENST00000398548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
ENST00000422911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
ENST00000433067.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
ENST00000441787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
ENST00000447016.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
ENST00000447207.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
ENST00000449395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
NM_001040424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
NM_001282934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
NM_022115.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
NR_104257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
NR_104258.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155727.hg.1 |
NR_104260.1 |
Refseq |