Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018766.hg.1 |
ENST00000344099.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018766.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018766.hg.1 |
TC19001308.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010078.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010078.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010078.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010078.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010078.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010078.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010078.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010078.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010078.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010078.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010078.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010078.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010078.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010078.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010078.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010078.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010078.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010078.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018767.hg.1 |
ENST00000344099.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018767.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018767.hg.1 |
TC19001308.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018764.hg.1 |
ENST00000344099.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018764.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018764.hg.1 |
TC19001308.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018765.hg.1 |
ENST00000344099.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018765.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018765.hg.1 |
TC19001308.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900183113.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900183113.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900183113.hg.1 |
TC1900010078.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900183113.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900183113.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900183116.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900183116.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900183116.hg.1 |
TC1900010078.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900183116.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900183116.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900183112.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900183112.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900183112.hg.1 |
TC1900010078.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900183112.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900183112.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010078.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010078.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010078.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010078.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010078.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010078.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010078.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010078.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010078.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010078.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010078.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010078.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010078.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010078.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010078.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010078.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010078.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010078.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900183115.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900183115.hg.1 |
NM_021030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900183115.hg.1 |
TC1900010078.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900183115.hg.1 |
ENST00000344099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900183115.hg.1 |
NM_021030.2 |
Refseq |