Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
ENST00000475682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
ENST00000475682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
ENST00000475682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
ENST00000475682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019028.hg.1 |
ENST00000341233.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019028.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019028.hg.1 |
ENST00000544887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019028.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019028.hg.1 |
TC10001506.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000177329.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177329.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177329.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177329.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177329.hg.1 |
TC1000011381.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
ENST00000371790.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
ENST00000475682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
ENST00000475682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
ENST00000475682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
ENST00000475682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011381.hg.1 |
NM_213606.3 |
Refseq |