Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006634.hg.1 |
TC09002907.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006634.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006634.hg.1 |
ENST00000477456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006634.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006635.hg.1 |
TC09002907.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006635.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006635.hg.1 |
ENST00000477456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006635.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184478.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184478.hg.1 |
ENST00000477456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184478.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184478.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184478.hg.1 |
ENST00000477456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184478.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184478.hg.1 |
TC0900012162.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900184479.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184479.hg.1 |
ENST00000477456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184479.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184479.hg.1 |
ENST00000259396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184479.hg.1 |
ENST00000477456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184479.hg.1 |
NM_000607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184479.hg.1 |
TC0900012162.hg.1 |
Clariom D TC |