Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIP5K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011522259.1 | XP_011520561.1 | 21 - 22 | 2631 | 101 | |
XM_017022738.1 | XP_016878227.1 | 20 - 21 | 2545 | 101 | ||
XM_017022745.1 | XP_016878234.1 | 21 - 22 | 2734 | 101 | ||
XM_017022746.1 | XP_016878235.1 | 20 - 21 | 2605 | 101 | ||
XM_017022752.1 | XP_016878241.1 | 21 - 22 | 2631 | 101 | ||
XM_017022753.1 | XP_016878242.1 | 20 - 21 | 2605 | 101 | ||
GenBank mRNA | AF502589.1 | - | 20 - 21 | 2553 | 101 | |
AF543190.1 | - | 20 - 21 | 2503 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |