Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173808.2 | NP_776169.2 | 5 - 6 | 884 | 107 | |
XM_011541200.2 | XP_011539502.1 | 5 - 6 | 1140 | 107 | ||
GenBank mRNA | AK092307.1 | - | 5 - 6 | 791 | 107 | |
AK295481.1 | - | 4 - 5 | 650 | 107 | ||
BC036771.1 | - | 5 - 6 | 806 | 5576828 | 107 | |
BX538014.1 | - | 5 - 6 | 788 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |