Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013209.hg.1 |
ENST00000426710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0M00006437.hg.1 |
ENST00000361739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0M00006437.hg.1 |
ENST00000361739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0M00006437.hg.1 |
ENST00000361739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0M00006437.hg.1 |
ENST00000361739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0M00006437.hg.1 |
ENST00000361739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0M00006437.hg.1 |
ENST00000361739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0M00006437.hg.1 |
ENST00000361739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0M00006437.hg.1 |
ENST00000361739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013209.hg.1 |
ENST00000426710.1 |
Ensembl |