Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008212.hg.1 |
ENST00000402176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008212.hg.1 |
ENST00000402176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008212.hg.1 |
ENST00000402176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008212.hg.1 |
ENST00000402176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008212.hg.1 |
ENST00000402176.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008212.hg.1 |
ENST00000402176.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008212.hg.1 |
ENST00000402176.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008212.hg.1 |
ENST00000402176.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008212.hg.1 |
ENST00000402176.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008212.hg.1 |
ENST00000402176.2 |
Ensembl |