Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006439.hg.1 |
ENST00000500893.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006439.hg.1 |
ENST00000500893.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006439.hg.1 |
ENST00000500893.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006439.hg.1 |
ENST00000500893.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006439.hg.1 |
ENST00000500893.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006439.hg.1 |
NM_033089.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006439.hg.1 |
NM_033089.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006439.hg.1 |
NM_033089.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006439.hg.1 |
NM_033089.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006439.hg.1 |
ENST00000500893.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006439.hg.1 |
ENST00000500893.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006439.hg.1 |
ENST00000500893.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006439.hg.1 |
ENST00000500893.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006439.hg.1 |
ENST00000500893.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006439.hg.1 |
NM_033089.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006439.hg.1 |
NM_033089.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006439.hg.1 |
NM_033089.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006439.hg.1 |
NM_033089.6 |
Refseq |