Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL109818.1 | - | 1 | 753 | 69 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chromosome 21 open reading frame 59 | C21orf59 |
C21orf48, CILD26, FBB18, Kur |
-31.4kb | Sense | Chr. 21 - 32601674 - 32612608 on NCBI Build GRCh38 |
Downstream | PAXBP1 antisense RNA 1 | PAXBP1-AS1 | GCFC1-AS1 | 82.4kb | Anti-sense | Chr. 21 - 32728114 - 32743122 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |