Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLSTN2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_108084.1 | 1 - 2 | 2143 | 80 | ||
GenBank mRNA | AL133103.1 | - | 1 - 2 | 2143 | 80 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | calsyntenin 2 | CLSTN2 |
ALC-GAMMA, CDHR13, CS2, CSTN2, alcagamma |
Anti-sense | Chr. 3 - 139935185 - 140577397 on NCBI Build GRCh38 | |
Downstream | tripartite motif containing 42 | TRIM42 |
PPP1R40, T4A1 |
169.2kb | Anti-sense | Chr. 3 - 140678024 - 140701150 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Probe spans exons |