Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cold shock domain containing E1 | CSDE1 |
D1S155E, UNR |
Sense | Chr. 1 - 114716913 - 114758050 on NCBI Build GRCh38 | |
Downstream | suppressor of IKBKE 1 | SIKE1 | SIKE | 27.4kb | Sense | Chr. 1 - 114769479 - 114780687 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |