Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLX1NB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_130724.1 | 3 | 918 | 108 | ||
GenBank mRNA | Y11609.1 | - | 1 | 147 | 107 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Kazal type serine peptidase inhibitor domain 1 | KAZALD1 |
BONO1, FKSG28, FKSG40, IGFBP-rP10 |
-46.7kb | Anti-sense | Chr. 10 - 101061141 - 101068141 on NCBI Build GRCh38 |
Downstream | T-cell leukemia homeobox 1 | TLX1 |
HOX11, TCL3 |
15.6kb | Anti-sense | Chr. 10 - 101130773 - 101137789 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |