Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
TATA-box binding protein associated factor, RNA polymerase I subunit B |
TAF1B |
MGC:9349, RAF1B, RAFI63, SL1, TAFI63 |
-2.5kb | Sense | Chr. 2 - 9843442 - 9934416 on NCBI Build GRCh38 |
Downstream | grainyhead like transcription factor 1 | GRHL1 |
LBP32, MGR, NH32, TFCP2L2 |
12.5kb | Sense | Chr. 2 - 9951663 - 10002284 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |