Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SAP domain containing ribonucleoprotein | SARNP |
CIP29, HCC1, HSPC316, THO1 |
Sense | Chr. 12 - 55752463 - 55817756 on NCBI Build GRCh38 | |
Downstream | ORMDL sphingolipid biosynthesis regulator 2 | ORMDL2 |
HSPC160, MST095, MSTP095, adoplin-2 |
46.9kb | Anti-sense | Chr. 12 - 55818022 - 55821175 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |