Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
secretoglobin family 1B member 2, pseudogene |
SCGB1B2P | SCGB4A1P | -45.2kb | Sense | Chr. 19 - 34576733 - 34677031 on NCBI Build GRCh38 |
Downstream |
secretoglobin family 2B member 3, pseudogene |
SCGB2B3P |
C2B, SCGB4A3P |
19.6kb | Anti-sense | Chr. 19 - 34645873 - 34646052 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |