Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC063630.1 | - | 1 | 655 | 77 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mitotic spindle positioning | MISP |
C19orf21, MISP1 |
-16.7kb | Anti-sense | Chr. 19 - 748413 - 764319 on NCBI Build GRCh38 |
Downstream | polypyrimidine tract binding protein 1 | PTBP1 |
HNRNP-I, HNRNPI, HNRPI, PTB, PTB-1, PTB-T, PTB2, PTB3, PTB4, pPTB |
15.5kb | Anti-sense | Chr. 19 - 797392 - 812327 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |