Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000289703.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000289703.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000289703.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000289703.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000289703.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000289703.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000289703.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000468523.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000468523.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000468523.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000617725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000617725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000617725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
ENST00000617725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_021795.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_021795.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_021795.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_021795.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_021795.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_021795.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_021795.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017072.hg.1 |
NM_021795.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229338.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229338.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229338.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229338.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229338.hg.1 |
TC0100017072.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100229339.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229339.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229339.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229339.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229339.hg.1 |
TC0100017072.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100229340.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229340.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229340.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229340.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229340.hg.1 |
TC0100017072.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000289703.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000289703.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000289703.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000289703.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000289703.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000289703.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000289703.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000357992.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000468523.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000468523.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000468523.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000616704.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000617725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000617725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000617725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
ENST00000617725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_001973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_021795.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_021795.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_021795.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_021795.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_021795.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_021795.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_021795.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017072.hg.1 |
NM_021795.2 |
Refseq |