Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000462.hg.1 |
TC01000038.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000462.hg.1 |
ENST00000378821.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000462.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000464.hg.1 |
TC01000038.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000464.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000463.hg.1 |
TC01000038.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000463.hg.1 |
ENST00000378821.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000463.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000461.hg.1 |
TC01000038.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000461.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006514.hg.1 |
ENST00000378821.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006514.hg.1 |
ENST00000378821.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006514.hg.1 |
ENST00000378821.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006514.hg.1 |
ENST00000378821.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006514.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006514.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006514.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006514.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144850.hg.1 |
ENST00000378821.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144850.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144850.hg.1 |
ENST00000378821.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144850.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144850.hg.1 |
TC0100006514.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100144853.hg.1 |
ENST00000378821.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144853.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144853.hg.1 |
ENST00000378821.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144853.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144853.hg.1 |
TC0100006514.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100144851.hg.1 |
ENST00000378821.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144851.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144851.hg.1 |
ENST00000378821.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144851.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144851.hg.1 |
TC0100006514.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100144852.hg.1 |
ENST00000378821.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144852.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144852.hg.1 |
ENST00000378821.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144852.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144852.hg.1 |
TC0100006514.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100006514.hg.1 |
ENST00000378821.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006514.hg.1 |
ENST00000378821.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006514.hg.1 |
ENST00000378821.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006514.hg.1 |
ENST00000378821.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006514.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006514.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006514.hg.1 |
NM_001146685.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006514.hg.1 |
NM_001146685.1 |
Refseq |