Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031133.hg.1 |
TC01002037.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031133.hg.1 |
ENST00000366474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031133.hg.1 |
NM_030904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012356.hg.1 |
ENST00000366474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012356.hg.1 |
NM_030904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031132.hg.1 |
TC01002037.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031132.hg.1 |
ENST00000366474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031132.hg.1 |
NM_030904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100190634.hg.1 |
ENST00000366474.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100190634.hg.1 |
NM_030904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100190634.hg.1 |
ENST00000366474.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100190634.hg.1 |
NM_030904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100190634.hg.1 |
TC0100012356.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100012356.hg.1 |
ENST00000366474.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012356.hg.1 |
NM_030904.1 |
Refseq |