Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057749.2 | NP_477097.1 | 9 - 10 | 925 | 103 | |
XM_017013957.1 | XP_016869446.1 | 8 - 9 | 1250 | 103 | ||
XM_017013958.1 | XP_016869447.1 | 9 - 10 | 1067 | 103 | ||
XM_017013959.1 | XP_016869448.1 | 9 - 10 | 1169 | 103 | ||
GenBank mRNA | AF091433.1 | - | 8 - 9 | 858 | 103 | |
AF102778.1 | - | 7 - 8 | 821 | 103 | ||
AF106690.1 | - | 7 - 8 | 821 | 103 | ||
AF112857.1 | - | 7 - 8 | 686 | 103 | ||
BC007015.1 | - | 7 - 8 | 660 | 3997513 | 103 | |
BC020729.1 | - | 9 - 10 | 872 | 4774554 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |