Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145873.1 | NP_001139345.1 | 9 | 2563 | 93 | |
NM_001768.6 | NP_001759.3 | 6 | 2421 | 93 | ||
NM_171827.3 | NP_741969.1 | 5 | 2310 | 93 | ||
NR_027353.1 | 7 | 2007 | 93 | |||
GenBank mRNA | AK097942.1 | - | 7 | 1605 | 93 | |
AK124156.1 | - | 9 | 2563 | 93 | ||
BC025715.1 | - | 6 | 1536 | 5227906 | 93 | |
M12824.1 | - | 6 | 1619 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |