Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGLL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182645.3 | NP_872586.1 | 3 - 4 | 1115 | 57 | |
GenBank mRNA | AJ578053.1 | - | 3 - 4 | 1099 | 57 | |
AK096878.1 | - | 3 - 4 | 932 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons |