Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012784.hg.1 |
ENST00000453129.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012784.hg.1 |
NM_001101389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012784.hg.1 |
TC11001020.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009091.hg.1 |
ENST00000453129.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009091.hg.1 |
ENST00000453129.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009091.hg.1 |
NM_001101389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100167418.hg.1 |
ENST00000453129.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167418.hg.1 |
NM_001101389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100167418.hg.1 |
ENST00000453129.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167418.hg.1 |
NM_001101389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100167418.hg.1 |
TC1100009091.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009091.hg.1 |
ENST00000453129.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009091.hg.1 |
ENST00000453129.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009091.hg.1 |
NM_001101389.1 |
Refseq |