Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101389.1 | NP_001094859.1 | 1 | 518 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |