Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006871.hg.1 |
ENST00000438745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000325307.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000325307.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000325307.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000325307.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000325307.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000325307.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000325307.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000419110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000419110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000419110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000419110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000419110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000419110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000430118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000430118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000430118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000430118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000430118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000448905.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000448905.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000448905.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000448905.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000448905.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000448905.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000455596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000455596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000455596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000455596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000455596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
ENST00000455596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_001301231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_005342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_005342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_005342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_005342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_005342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_005342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008724.hg.1 |
NM_005342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009983.hg.1 |
ENST00000433260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009983.hg.1 |
ENST00000433260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009701.hg.1 |
ENST00000442010.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009701.hg.1 |
ENST00000442010.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009701.hg.1 |
ENST00000451823.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009701.hg.1 |
ENST00000451823.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009701.hg.1 |
ENST00000451823.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008004.hg.1 |
ENST00000567416.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008887.hg.1 |
ENST00000511263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008887.hg.1 |
ENST00000511263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008887.hg.1 |
ENST00000511263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008887.hg.1 |
ENST00000581508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008887.hg.1 |
NR_039951.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009185.hg.1 |
ENST00000434734.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009185.hg.1 |
ENST00000434734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009185.hg.1 |
ENST00000434734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009185.hg.1 |
ENST00000434734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009701.hg.1 |
ENST00000442010.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009701.hg.1 |
ENST00000442010.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009701.hg.1 |
ENST00000451823.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009701.hg.1 |
ENST00000451823.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009701.hg.1 |
ENST00000451823.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008004.hg.1 |
ENST00000567416.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006871.hg.1 |
ENST00000438745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008887.hg.1 |
ENST00000511263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008887.hg.1 |
ENST00000511263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008887.hg.1 |
ENST00000511263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008887.hg.1 |
ENST00000581508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008887.hg.1 |
NR_039951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009983.hg.1 |
ENST00000433260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009983.hg.1 |
ENST00000433260.1 |
Ensembl |