Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033557.2 | NP_001028729.1 | 2 - 3 | 1331 | 98 | |
NM_002706.5 | NP_002697.1 | 2 - 3 | 1331 | 98 | ||
NM_177968.3 | NP_808907.1 | 2 - 3 | 1331 | 98 | ||
XM_011532936.2 | XP_011531238.1 | 2 - 3 | 1289 | 98 | ||
XM_017004395.1 | XP_016859884.1 | 2 - 3 | 1331 | 98 | ||
XM_017004396.1 | XP_016859885.1 | 2 - 3 | 1331 | 98 | ||
XM_017004397.1 | XP_016859886.1 | 2 - 3 | 1331 | 98 | ||
GenBank mRNA | AF136972.1 | - | 1 - 2 | 845 | 98 | |
AF294792.1 | - | 1 - 2 | 845 | 98 | ||
AJ005801.1 | - | 1 - 2 | 845 | 98 | ||
AJ271832.1 | - | 2 - 3 | 1238 | 98 | ||
AJ271835.1 | - | 2 - 3 | 1238 | 98 | ||
AK056009.1 | - | 2 - 3 | 1267 | 98 | ||
AK291214.1 | - | 2 - 3 | 1273 | 98 | ||
AL833035.1 | - | 2 - 3 | 1136 | 98 | ||
BC064381.1 | - | 2 - 3 | 1246 | 5734287 | 98 | |
DQ023508.1 | - | 2 - 3 | 1257 | 98 | ||
DQ023509.1 | - | 2 - 3 | 1257 | 98 | ||
DQ023510.1 | - | 2 - 3 | 1257 | 98 | ||
DQ023511.1 | - | 2 - 3 | 1257 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Homo sapiens Probe spans exons |