Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR8068 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_107035.1 | 1 | 31 | 67 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | methyltransferase like 15 | METTL15 | METT5D1 | Anti-sense | Chr. 11 - 28108251 - 28527041 on NCBI Build GRCh38 | |
Downstream | potassium voltage-gated channel subfamily A member 4 | KCNA4 |
HBK4, HK1, HPCN2, HUKII, KCNA4L, KCNA8, KV1.4, PCN2 |
1.5mb | Sense | Chr. 11 - 30009741 - 30017030 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |