Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199640.1 | NP_001186569.1 | 5 - 6 | 474 | 83 | |
NM_001199641.1 | NP_001186570.1 | 3 - 4 | 222 | 83 | ||
NM_001314044.1 | NP_001300973.1 | 6 - 7 | 587 | 83 | ||
NM_001314045.1 | NP_001300974.1 | 6 - 7 | 595 | 83 | ||
NM_001314046.1 | NP_001300975.1 | 6 - 7 | 582 | 83 | ||
NM_001314047.1 | NP_001300976.1 | 6 - 7 | 569 | 83 | ||
NM_001314048.1 | NP_001300977.1 | 5 - 6 | 474 | 83 | ||
NM_033439.3 | NP_254274.1 | 6 - 7 | 600 | 83 | ||
XM_011518061.2 | XP_011516363.1 | 5 - 6 | 535 | 83 | ||
XM_017015285.1 | XP_016870774.1 | 8 - 9 | 1328 | 83 | ||
GenBank mRNA | AB024518.1 | - | 5 - 6 | 539 | 83 | |
AK295908.1 | - | 3 - 4 | 203 | 83 | ||
AK303943.1 | - | 5 - 6 | 451 | 83 | ||
AK313414.1 | - | 6 - 7 | 579 | 83 | ||
AY905581.1 | - | 5 - 6 | 522 | 83 | ||
BC047085.1 | - | 6 - 7 | 577 | 5314040 | 83 | |
BQ878541.1 | - | 5 - 6 | 561 | 83 | ||
CR407619.1 | - | 5 - 6 | 522 | 83 | ||
HQ641439.1 | - | 4 - 5 | 411 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons |