Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283747.1 | NP_001270676.1 | 3 - 4 | 539 | 83 | |
GenBank mRNA | AB169816.1 | - | 3 - 4 | 539 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |