Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005568315.2 | XP_005568372.1 | 2 - 3 | 560 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |