Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005587809.2 | XP_005587866.1 | 2 - 3 | 363 | 110 | |
XM_005587810.2 | XP_005587867.1 | 2 - 3 | 305 | 110 | ||
GenBank mRNA | AB169436.1 | - | 2 - 3 | 160 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |