Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005582850.2 | XP_005582907.1 | 3 - 4 | 1014 | 154 | |
XM_005582851.2 | XP_005582908.1 | 3 - 4 | 942 | 154 | ||
XM_005582853.2 | XP_005582910.1 | 2 - 3 | 768 | 154 | ||
XM_005582854.2 | XP_005582911.1 | 4 - 5 | 603 | 154 | ||
XM_005582855.2 | XP_005582912.1 | 3 - 4 | 708 | 154 | ||
XM_005582856.2 | XP_005582913.1 | 3 - 4 | 636 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |