Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284952.1 | NP_001271881.1 | 3 - 4 | 324 | 86 | |
GenBank mRNA | AB172966.1 | - | 3 - 4 | 324 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Probe spans exons |