Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287327.1 | NP_001274256.1 | 5 - 6 | 425 | 96 | |
XM_005550298.2 | XP_005550355.1 | 5 - 6 | 436 | 96 | ||
GenBank mRNA | AB173436.1 | - | 5 - 6 | 425 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |